Product Details
- SNP ID
-
rs4513569
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:7049905 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCGCTAGCTGCAGGTGGCCCTAGT[C/T]GTTGGCCCAGTGTCTGAGGCTGAGC
- Phenotype
-
MIM: 606173
MIM: 608790
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC96
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115659921] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC96
- Gene Name
- coiled-coil domain containing 96
There are no transcripts associated with this gene.
- Gene
- GRPEL1
- Gene Name
- GrpE like 1, mitochondrial
There are no transcripts associated with this gene.
- Gene
- LOC100129931
- Gene Name
- uncharacterized LOC100129931
There are no transcripts associated with this gene.
- Gene
- TADA2B
- Gene Name
- transcriptional adaptor 2B
View Full Product Details