Product Details
- SNP ID
-
rs4750017
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:6146045 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTCCTGCGCCGTCTGTTTGGGTT[A/G]GGAGGGGTTGAGTTCTGGTGATGAT
- Phenotype
-
MIM: 605319
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3155A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150280344] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR3155A
- Gene Name
- microRNA 3155a
There are no transcripts associated with this gene.
- Gene
- MIR3155B
- Gene Name
- microRNA 3155b
There are no transcripts associated with this gene.
- Gene
- PFKFB3
- Gene Name
- 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
View Full Product Details