Product Details

SNP ID
hCV27978277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:127948451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATCTTGGTTCACTCTGATTTCT[C/G]TATGCCAGACTAGTAGAGCTTCAGG
Phenotype
MIM: 165140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PVT1 PubMed Links

Gene Details

Gene
PVT1
Gene Name
Pvt1 oncogene (non-protein coding)
There are no transcripts associated with this gene.

Gene
TMEM75
Gene Name
transmembrane protein 75
There are no transcripts associated with this gene.

View Full Product Details