Product Details

SNP ID
rs4827380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:66597898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAATCTGGCTCCCCAGACTACAA[A/C]AGGGAAGCAAGAAATGCTCCAGATC
Phenotype
MIM: 300276
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EDA2R PubMed Links

Gene Details

Gene
EDA2R
Gene Name
ectodysplasin A2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199687.2 1271 UTR 3 NP_001186616.1
NM_001242310.1 1271 UTR 3 NP_001229239.1
NM_001324199.1 1271 UTR 3 NP_001311128.1
NM_001324201.1 1271 UTR 3 NP_001311130.1
NM_001324202.1 1271 UTR 3 NP_001311131.1
NM_001324204.1 1271 UTR 3 NP_001311133.1
NM_001324205.1 1271 UTR 3 NP_001311134.1
NM_001324206.1 1271 UTR 3 NP_001311135.1
NM_021783.4 1271 UTR 3 NP_068555.1
XM_011531000.1 1271 UTR 3 XP_011529302.1
XM_011531001.1 1271 UTR 3 XP_011529303.1
XM_011531002.1 1271 UTR 3 XP_011529304.1
XM_011531003.2 1271 UTR 3 XP_011529305.1
XM_011531004.1 1271 Intron XP_011529306.1
XM_011531005.2 1271 UTR 3 XP_011529307.1
XM_011531006.2 1271 UTR 3 XP_011529308.1
XM_011531007.2 1271 Intron XP_011529309.1
XM_017029703.1 1271 UTR 3 XP_016885192.1
XM_017029704.1 1271 UTR 3 XP_016885193.1
XM_017029705.1 1271 UTR 3 XP_016885194.1
XM_017029706.1 1271 UTR 3 XP_016885195.1
XM_017029707.1 1271 UTR 3 XP_016885196.1
XM_017029708.1 1271 UTR 3 XP_016885197.1

View Full Product Details