Product Details

SNP ID
rs4984805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1309216 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCCTTAGGCCTCAGTGGTCTTTG[A/G]CCCCCGGCCCCAGGACCTGACCCCA
Phenotype
MIM: 601661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2I PubMed Links

Gene Details

Gene
UBE2I
Gene Name
ubiquitin conjugating enzyme E2 I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003345.4 46 Intron NP_003336.1
NM_194259.2 46 Intron NP_919235.1
NM_194260.2 46 UTR 5 NP_919236.1
NM_194261.2 46 UTR 5 NP_919237.1
XM_005255540.2 46 Intron XP_005255597.1
XM_005255541.4 46 Intron XP_005255598.1
XM_011522644.2 46 Intron XP_011520946.1
XM_011522645.1 46 UTR 5 XP_011520947.1
XM_017023640.1 46 Intron XP_016879129.1

View Full Product Details