Product Details

SNP ID
rs4523234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143040059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCTCCTTGGGCTGAACCCTCT[A/G]TCTAGGGGACAATGGCCCTGGGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8orf31 PubMed Links

Gene Details

Gene
C8orf31
Gene Name
chromosome 8 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173687.3 Intron NP_775958.1
XM_017013344.1 Intron XP_016868833.1
XM_017013345.1 Intron XP_016868834.1
XM_017013346.1 Intron XP_016868835.1
Gene
LOC107984097
Gene Name
uncharacterized protein FLJ40521
There are no transcripts associated with this gene.

View Full Product Details