Product Details

SNP ID
rs4467241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171526929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATCTCAAGTTGATTTCGTGAGA[G/T]AATATAACTACCTTTAAGTAACATT
Phenotype
MIM: 605745
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYBRD1 PubMed Links

Gene Details

Gene
CYBRD1
Gene Name
cytochrome b reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127383.1 Intron NP_001120855.1
NM_001256909.1 Intron NP_001243838.1
NM_024843.3 Intron NP_079119.3

View Full Product Details