Product Details

SNP ID
rs4723716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:37921353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGATCTTTCGCGAGGAGGGTC[G/T]CTTGGGGATAGCATGGTCCGGGACT
Phenotype
MIM: 606570
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EPDR1 PubMed Links

Gene Details

Gene
EPDR1
Gene Name
ependymin related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242946.1 35 Intron NP_001229875.2
NM_001242948.1 35 UTR 5 NP_001229877.1
NM_017549.4 35 Intron NP_060019.2
Gene
SFRP4
Gene Name
secreted frizzled related protein 4
There are no transcripts associated with this gene.

View Full Product Details