Product Details

SNP ID
rs4801801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49488371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTAAGTATAAAGGAGGGATTGG[C/T]CCTGGAAAGGTTTCTTTGTGAGTAG
Phenotype
MIM: 600007 MIM: 180471
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLT3LG PubMed Links

Gene Details

Gene
FLT3LG
Gene Name
fms related tyrosine kinase 3 ligand
There are no transcripts associated with this gene.

Gene
RPL13A
Gene Name
ribosomal protein L13a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270491.1 Intron NP_001257420.1
NM_012423.3 Intron NP_036555.1
Gene
RPS11
Gene Name
ribosomal protein S11
There are no transcripts associated with this gene.

Gene
SNORD32A
Gene Name
small nucleolar RNA, C/D box 32A
There are no transcripts associated with this gene.

Gene
SNORD33
Gene Name
small nucleolar RNA, C/D box 33
There are no transcripts associated with this gene.

Gene
SNORD34
Gene Name
small nucleolar RNA, C/D box 34
There are no transcripts associated with this gene.

Gene
SNORD35A
Gene Name
small nucleolar RNA, C/D box 35A
There are no transcripts associated with this gene.

Gene
SNORD35B
Gene Name
small nucleolar RNA, C/D box 35B
There are no transcripts associated with this gene.

View Full Product Details