Product Details
- SNP ID
-
rs4233875
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:10069971 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAAGTCAGAGCTGCCTGTGAGGGA[A/G]AGGCTGCCCCAGGAGGCAGTGAGAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYS1
PubMed Links
Gene Details
- Gene
- CYS1
- Gene Name
- cystin 1
View Full Product Details