Product Details

SNP ID
rs4393538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:33315205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGAGCTGCAGCGCTCACGTGAG[A/C]TCATGTCTCTCGTCCCTCCCCAGAG
Phenotype
MIM: 180903
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928134 PubMed Links

Gene Details

Gene
LOC101928134
Gene Name
uncharacterized LOC101928134
There are no transcripts associated with this gene.

Gene
RYR3
Gene Name
ryanodine receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001036.4 Intron NP_001027.3
NM_001243996.2 Intron NP_001230925.1

View Full Product Details