Product Details
- SNP ID
-
rs4576654
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:156927807 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCATCCAGGACAAGGGCGCCCTGAA[A/G]CTGGCTGAGGTGGGTGTGCCGATCA
- Phenotype
-
MIM: 605708
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF11
PubMed Links
Gene Details
- Gene
- ARHGEF11
- Gene Name
- Rho guanine nucleotide exchange factor 11
There are no transcripts associated with this gene.
- Gene
- LRRC71
- Gene Name
- leucine rich repeat containing 71
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144702.2 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
NP_653303.2 |
XM_005244926.3 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
XP_005244983.1 |
XM_005244927.3 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
XP_005244984.1 |
XM_005244928.2 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
XP_005244985.1 |
XM_006711185.3 |
1401 |
Silent Mutation |
AAA,AAG |
K272K |
XP_006711248.1 |
XM_006711186.3 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
XP_006711249.1 |
XM_006711187.3 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
XP_006711250.1 |
XM_011509239.2 |
1401 |
Silent Mutation |
AAA,AAG |
K113K |
XP_011507541.1 |
XM_011509240.2 |
1401 |
Silent Mutation |
AAA,AAG |
K84K |
XP_011507542.1 |
XM_011509241.2 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
XP_011507543.1 |
XM_017000459.1 |
1401 |
Silent Mutation |
AAA,AAG |
K299K |
XP_016855948.1 |
XM_017000460.1 |
1401 |
Silent Mutation |
AAA,AAG |
K84K |
XP_016855949.1 |
XM_017000461.1 |
1401 |
Silent Mutation |
AAA,AAG |
K84K |
XP_016855950.1 |
XM_017000462.1 |
1401 |
Silent Mutation |
AAA,AAG |
K84K |
XP_016855951.1 |
- Gene
- MIR765
- Gene Name
- microRNA 765
There are no transcripts associated with this gene.
View Full Product Details