Product Details

SNP ID
rs4305200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15382874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTAAAGAAAAAGGTTTTGGATTT[C/T]GGAGCATTTCGGATTTTCAGATGAC
Phenotype
MIM: 604692 MIM: 609475
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP8 PubMed Links

Gene Details

Gene
AKAP8
Gene Name
A-kinase anchoring protein 8
There are no transcripts associated with this gene.

Gene
AKAP8L
Gene Name
A-kinase anchoring protein 8 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291478.1 Intron NP_001278407.1
NM_014371.3 Intron NP_055186.3
XM_005259854.3 Intron XP_005259911.2
XM_011527916.2 Intron XP_011526218.1

View Full Product Details