Product Details

SNP ID
rs4599403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:84967482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAATTTGGGTCACGCTTCAGTT[C/T]TGGCAGTAAAGACAAACTGAGTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDFY3 PubMed Links

Gene Details

Gene
WDFY3
Gene Name
WD repeat and FYVE domain containing 3
There are no transcripts associated with this gene.

Gene
WDFY3-AS2
Gene Name
WDFY3 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details