Product Details

SNP ID
rs4942267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44028144 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTATGCTTCCAAAATTCACTCTCA[C/T]TCTTTGTATGAGAGGCCAATTAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00284 PubMed Links

Gene Details

Gene
LINC00284
Gene Name
long intergenic non-protein coding RNA 284
There are no transcripts associated with this gene.

View Full Product Details