Product Details
- SNP ID
-
rs4351605
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:32751169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCAGCTCATTCTTGTGGTTCCTG[A/G]AGTTCTGGTCCATATGCCTTCCATA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1522
PubMed Links
Gene Details
- Gene
- KIAA1522
- Gene Name
- KIAA1522
View Full Product Details