Product Details
- SNP ID
-
rs4235825
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:85457325 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTACTTCTTGGCTCTTCCACTTA[C/T]TAGCTGTGTGAATTTGGCAGTGGTT
- Phenotype
-
MIM: 129190
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NT5E
PubMed Links
Gene Details
- Gene
- NT5E
- Gene Name
- 5'-nucleotidase ecto
View Full Product Details