Product Details

SNP ID
rs4384501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:69702068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATCAAAGCTCTATTCTGGAGTT[G/T]GCATGGACACATTTGTGATAATATT
Phenotype
MIM: 605332
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL1 PubMed Links

Gene Details

Gene
KLHL1
Gene Name
kelch like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286725.1 Intron NP_001273654.1
NM_020866.2 Intron NP_065917.1

View Full Product Details