Product Details

SNP ID
rs4809526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63298665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCATTCAGAACAGCCTGGACTCC[A/G]CACAGCGAACCCCAGAAGCACCTGC
Phenotype
MIM: 608377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFGAP1 PubMed Links

Gene Details

Gene
ARFGAP1
Gene Name
ADP ribosylation factor GTPase activating protein 1
There are no transcripts associated with this gene.

Gene
COL20A1
Gene Name
collagen type XX alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020882.2 Intron NP_065933.2
XM_011528937.1 Intron XP_011527239.1
XM_011528938.1 Intron XP_011527240.1
XM_011528939.1 Intron XP_011527241.1
XM_011528940.1 Intron XP_011527242.1

View Full Product Details