Product Details

SNP ID
rs4417283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:122885302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAACTATGGAAAACTACACTCTT[C/T]TTTCTTCTTACTTTACTTTCCAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf63 PubMed Links

Gene Details

Gene
C11orf63
Gene Name
chromosome 11 open reading frame 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024806.3 Intron NP_079082.2
NM_199124.2 Intron NP_954575.1

View Full Product Details