Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001038618.2 | Intron | NP_001033707.1 | |||
NM_001083608.1 | Intron | NP_001077077.1 | |||
NM_012336.3 | Intron | NP_036468.1 | |||
NM_031968.2 | Intron | NP_114174.1 | |||
XM_005256340.3 | Intron | XP_005256397.1 | |||
XM_006722278.3 | Intron | XP_006722341.1 | |||
XM_011523541.2 | Intron | XP_011521843.1 | |||
XM_011523543.2 | Intron | XP_011521845.1 | |||
XM_011523544.2 | Intron | XP_011521846.1 | |||
XM_017024449.1 | Intron | XP_016879938.1 | |||
XM_017024450.1 | Intron | XP_016879939.1 |