Product Details

SNP ID
rs4236546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100950265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGCTGTTGGTACCAGGGCCGGG[A/G]CAGGGAGCTCCTGATGTCCACTTCG
Phenotype
MIM: 158371
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUC3A PubMed Links

Gene Details

Gene
MUC3A
Gene Name
mucin 3A, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005960.1 Intron NP_005951.1
XM_011516246.2 Intron XP_011514548.1
XM_011516247.2 Intron XP_011514549.1
XM_011516249.2 Intron XP_011514551.1
XM_011516250.2 Intron XP_011514552.1
XM_017012231.1 Intron XP_016867720.1

View Full Product Details