Product Details

SNP ID
rs4917116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:50101472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCCCCAACTATCTTCTGAATCC[C/T]GGGAGAGAATAAATGGTTGACTTAA
Phenotype
MIM: 608498
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf72 PubMed Links

Gene Details

Gene
C7orf72
Gene Name
chromosome 7 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161834.2 Intron NP_001155306.2
XM_011515052.1 Intron XP_011513354.1
XM_011515053.2 Intron XP_011513355.1
Gene
ZPBP
Gene Name
zona pellucida binding protein
There are no transcripts associated with this gene.

View Full Product Details