Product Details
- SNP ID
-
rs4458916
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:11982922 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGAGAGGTCAGAATTTACATTCT[C/G]TGGACAGAAGCCAGCAACTTGGTGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB135
PubMed Links
Gene Details
- Gene
- DEFB135
- Gene Name
- defensin beta 135
- Gene
- DEFB136
- Gene Name
- defensin beta 136
There are no transcripts associated with this gene.
View Full Product Details