Product Details

SNP ID
rs4509937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99609347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCTCTGGGGAGGGGAGCCACG[C/T]TTCCTTTCCCATCCTCTGCACCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC85C PubMed Links

Gene Details

Gene
CCDC85C
Gene Name
coiled-coil domain containing 85C
There are no transcripts associated with this gene.

Gene
HHIPL1
Gene Name
HHIP like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006720277.3 Intron XP_006720340.1
XM_011537236.2 Intron XP_011535538.1
XM_011537237.2 Intron XP_011535539.1
XM_017021707.1 Intron XP_016877196.1
XM_017021708.1 Intron XP_016877197.1

View Full Product Details