Product Details

SNP ID
rs4276219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196643478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGCCCCAACCCAAAAGGTTG[C/T]CTAAGGCAGGTGCTGCGCCATGTGC
Phenotype
MIM: 615322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRROS PubMed Links

Gene Details

Gene
NRROS
Gene Name
negative regulator of reactive oxygen species
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198565.2 Intron NP_940967.1

View Full Product Details