Product Details

SNP ID
rs4491965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1214681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTGGCTGAGGATCTGCCCTGT[A/T]TGGACCTGCACGGCCCCACAGCTCC
Phenotype
MIM: 602618 MIM: 605918
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CTBP1 PubMed Links

Gene Details

Gene
CTBP1
Gene Name
C-terminal binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012614.1 Intron NP_001012632.1
NM_001328.2 Intron NP_001319.1
Gene
CTBP1-AS
Gene Name
CTBP1 antisense RNA
There are no transcripts associated with this gene.

Gene
LOC100130872
Gene Name
uncharacterized LOC100130872
There are no transcripts associated with this gene.

Gene
SPON2
Gene Name
spondin 2
There are no transcripts associated with this gene.

View Full Product Details