Product Details

SNP ID
rs4919643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102462620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTACCTACCAGGGGCCTGGTA[C/G]GGTGAAGGGGAAGAGTTTTGACACC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MFSD13A PubMed Links

Gene Details

Gene
MFSD13A
Gene Name
major facilitator superfamily domain containing 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024789.3 Intron NP_079065.2
XM_006717973.3 Intron XP_006718036.1
XM_006717974.3 Intron XP_006718037.1
XM_011540160.2 Intron XP_011538462.1
XM_011540163.2 Intron XP_011538465.1
XM_011540167.2 Intron XP_011538469.1
XM_011540169.2 Intron XP_011538471.1
XM_017016654.1 Intron XP_016872143.1
XM_017016655.1 Intron XP_016872144.1
XM_017016656.1 Intron XP_016872145.1
XM_017016657.1 Intron XP_016872146.1
XM_017016658.1 Intron XP_016872147.1
XM_017016659.1 Intron XP_016872148.1
XM_017016660.1 Intron XP_016872149.1
XM_017016661.1 Intron XP_016872150.1
XM_017016662.1 Intron XP_016872151.1
Gene
RPARP-AS1
Gene Name
RPARP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details