Product Details

SNP ID
rs4958278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151661323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATTTACTTCAATTCCTCTCTC[C/T]ACAGATTCCTAGAGATTATGAGACT
Phenotype
MIM: 182120
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPARC PubMed Links

Gene Details

Gene
SPARC
Gene Name
secreted protein acidic and cysteine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309443.1 3363 UTR 3 NP_001296372.1
NM_001309444.1 3363 UTR 3 NP_001296373.1
NM_003118.3 3363 UTR 3 NP_003109.1

View Full Product Details