Product Details

SNP ID
rs4881734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1477149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTATGAGGCTCCATCACTGAGG[A/G]GAGGGCTGCAATCCCCGGCGATCAT
Phenotype
MIM: 611969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOB2 PubMed Links

Gene Details

Gene
MOB2
Gene Name
MOB kinase activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172223.2 Intron NP_001165694.1
NM_053005.5 Intron NP_443731.2

View Full Product Details