Product Details
- SNP ID
-
rs4274064
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:206695009 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCTTTCCAGCAGAGCCCACCTG[C/G]GACTCTTAGTGACTTCATACCCTTT
- Phenotype
-
MIM: 602006
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MAPKAPK2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114787488] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAPKAPK2
- Gene Name
- mitogen-activated protein kinase-activated protein kinase 2
View Full Product Details