Product Details

SNP ID
rs4806878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2846784 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTTCCTAAGCGCTTTTGGTCTC[A/G]TGAATCTTAGAGTCCAAATTGACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF555 PubMed Links

Gene Details

Gene
ZNF555
Gene Name
zinc finger protein 555
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172775.1 Intron NP_001166246.1
NM_152791.4 Intron NP_690004.4
XM_011527716.2 Intron XP_011526018.1
XM_017026375.1 Intron XP_016881864.1

View Full Product Details