Product Details

SNP ID
rs4318447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57393563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCTGCCTTGGTACACACCCCA[A/G]CTCCCTTCGCTAGGTGTGTGGCCTC
Phenotype
MIM: 612428
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100291105 PubMed Links

Gene Details

Gene
LOC100291105
Gene Name
uncharacterized LOC100291105
There are no transcripts associated with this gene.

Gene
RBM38
Gene Name
RNA binding motif protein 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291780.1 Intron NP_001278709.1
NM_017495.5 Intron NP_059965.2
NM_183425.2 Intron NP_906270.1

View Full Product Details