Product Details

SNP ID
rs4435288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76873307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTCCTTGAGGCACACATTTCC[C/T]TGCCTTTGGCCCAGGCTGTGGCCAT
Phenotype
MIM: 612441
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGAT5B PubMed Links

Gene Details

Gene
MGAT5B
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199172.1 Intron NP_001186101.1
NM_144677.2 Intron NP_653278.2
NM_198955.1 Intron NP_945193.1

View Full Product Details