Product Details

SNP ID
rs4571441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:105099675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATTGGGACACAGCAGGTCAGGAG[C/T]GATTCTGAAGCCTGAGGATGCCATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB9BP1 PubMed Links

Gene Details

Gene
RAB9BP1
Gene Name
RAB9B, member RAS oncogene family pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details