Product Details

SNP ID
rs4312309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87950991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAACTACAAGTCCCAGCAACCCT[C/T]GGTGCGGGGACAGGACGGGCCACCG
Phenotype
MIM: 611564
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BANP PubMed Links

Gene Details

Gene
BANP
Gene Name
BTG3 associated nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173539.1 Intron NP_001167010.1
NM_001173540.1 Intron NP_001167011.1
NM_001173541.1 Intron NP_001167012.1
NM_001173542.1 Intron NP_001167013.1
NM_001173543.1 Intron NP_001167014.1
NM_017869.3 Intron NP_060339.2
NM_079837.2 Intron NP_524576.2
XM_005256008.3 Intron XP_005256065.1
XM_005256009.3 Intron XP_005256066.1
XM_011523170.2 Intron XP_011521472.1
XM_011523172.2 Intron XP_011521474.1
XM_011523174.2 Intron XP_011521476.1
XM_011523175.2 Intron XP_011521477.1
XM_011523176.2 Intron XP_011521478.1
XM_011523177.2 Intron XP_011521479.1
XM_011523178.2 Intron XP_011521480.1
XM_011523179.2 Intron XP_011521481.1
XM_011523180.2 Intron XP_011521482.1
XM_011523181.2 Intron XP_011521483.1
XM_011523182.1 Intron XP_011521484.1
XM_011523183.2 Intron XP_011521485.1
XM_011523184.2 Intron XP_011521486.1
XM_011523186.2 Intron XP_011521488.1
XM_017023378.1 Intron XP_016878867.1
XM_017023379.1 Intron XP_016878868.1
XM_017023380.1 Intron XP_016878869.1
XM_017023381.1 Intron XP_016878870.1
XM_017023382.1 Intron XP_016878871.1
XM_017023383.1 Intron XP_016878872.1

View Full Product Details