Product Details

SNP ID
rs4660734
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43526662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATCCCACCCTGACCTGCCCCCA[A/G]AAGAAAGTGGCTGCATGCGATTCGA
Phenotype
MIM: 179590
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPRF PubMed Links

Gene Details

Gene
PTPRF
Gene Name
protein tyrosine phosphatase, receptor type F
There are no transcripts associated with this gene.

View Full Product Details