Product Details
- SNP ID
-
hCV28032675
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:33554615 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCAAAAACCCTGGGCAATCCCAG[C/G]TGGGGACCTAGTACCAACCAGTCTG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RNF122
PubMed Links
Gene Details
- Gene
- RNF122
- Gene Name
- ring finger protein 122
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024787.3 |
|
Intron |
|
|
NP_079063.2 |
View Full Product Details