Product Details
- SNP ID
-
rs4922508
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:47323706 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGGGAAATAAGTTTCTGAAGAGC[G/A]AAACTGTTCAGTCAAAGCTTGCAGC
- Phenotype
-
MIM: 605120
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GDF2
PubMed Links
Gene Details
- Gene
- GDF2
- Gene Name
- growth differentiation factor 2
There are no transcripts associated with this gene.
View Full Product Details