Product Details

SNP ID
rs4407401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48752420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCAGCATACTCTACACAATTA[C/G]TGAGGAATTCACTTTGCAGAAAATT
Phenotype
MIM: 176910
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRKAR2A PubMed Links

Gene Details

Gene
PRKAR2A
Gene Name
protein kinase cAMP-dependent type II regulatory subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321982.1 Intron NP_001308911.1
NM_001321983.1 Intron NP_001308912.1
NM_001321989.1 Intron NP_001308918.1
NM_004157.3 Intron NP_004148.1

View Full Product Details