Product Details
- SNP ID
-
rs4495556
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:96239249 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGGGCATCCTCTCCCAGCGCCAC[C/T]TAGTGAGAGGGGCAGGAAGAAGGGA
- Phenotype
-
MIM: 605573
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSD17B3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74391725] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSD17B3
- Gene Name
- hydroxysteroid 17-beta dehydrogenase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000197.1 |
|
Intron |
|
|
NP_000188.1 |
View Full Product Details