Product Details

SNP ID
rs4395475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:46740632 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGTAATTGTTAGGTGTAAAATGGA[C/T]GGAGAGATGACAAAAATGCCAAAAA
Phenotype
MIM: 609811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX7B2 PubMed Links

Gene Details

Gene
COX7B2
Gene Name
cytochrome c oxidase subunit 7B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130902.2 Intron NP_570972.2
XM_005248056.4 Intron XP_005248113.1
XM_011513630.2 Intron XP_011511932.1
XM_011513631.2 Intron XP_011511933.1
XM_011513632.2 Intron XP_011511934.1
XM_011513633.2 Intron XP_011511935.1
XM_011513634.2 Intron XP_011511936.1
XM_011513635.2 Intron XP_011511937.1
XM_011513636.1 Intron XP_011511938.1
XM_011513637.2 Intron XP_011511939.1
XM_011513638.2 Intron XP_011511940.1

View Full Product Details