Product Details

SNP ID
rs4846057
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:9852836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCCTCTGTAGTGAGCTGCAGG[C/T]ACAAAAATAGCCCTGGATGCCCCAG
Phenotype
MIM: 607758
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNBIP1 PubMed Links

Gene Details

Gene
CTNNBIP1
Gene Name
catenin beta interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012329.1 Intron NP_001012329.1
NM_020248.2 Intron NP_064633.1

View Full Product Details