Product Details

SNP ID
rs7688908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152793005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACTTAATGACATTTTGGCCTCCA[A/G]TATAACTGGTTTGTGAGGGCTGAAC
Phenotype
MIM: 605928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFIP1 PubMed Links

Gene Details

Gene
ARFIP1
Gene Name
ADP ribosylation factor interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025593.2 Intron NP_001020764.1
NM_001025595.2 Intron NP_001020766.1
NM_001287431.1 Intron NP_001274360.1
NM_001287432.1 Intron NP_001274361.1
NM_001287433.1 Intron NP_001274362.1
NM_014447.3 Intron NP_055262.1
XM_011531868.2 Intron XP_011530170.1
XM_011531869.2 Intron XP_011530171.1
XM_011531870.2 Intron XP_011530172.1
XM_011531871.2 Intron XP_011530173.1
XM_011531872.1 Intron XP_011530174.1

View Full Product Details