Product Details
- SNP ID
-
rs8068646
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:59621459 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGACCATTTTTATTTACATTTCCC[C/T]ATTGGCATAGCATACATAAAGGATT
- Phenotype
-
MIM: 118955
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLTC
PubMed Links
Gene Details
- Gene
- CLTC
- Gene Name
- clathrin heavy chain
View Full Product Details