Product Details
- SNP ID
-
rs7612632
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:150974511 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTCAGCATAAATGAGTCTCTAC[A/G]TTATGCACACTTCGAAGGGAACTTT
- Phenotype
-
MIM: 606397
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLRN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143256983] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLRN1
- Gene Name
- clarin 1
There are no transcripts associated with this gene.
- Gene
- CLRN1-AS1
- Gene Name
- CLRN1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details