Product Details

SNP ID
rs6863930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177332690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGCTGGATGCTCTCGGCAGCCCC[C/T]CAGGAGGGTCTCCCAGGACCTGAGT
Phenotype
MIM: 609551
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMAN2 PubMed Links

Gene Details

Gene
LMAN2
Gene Name
lectin, mannose binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006816.2 Intron NP_006807.1

View Full Product Details