Product Details
- SNP ID
-
rs8083455
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:58682880 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTAAAATAGGCCTCTGATAAATGC[C/G]CTATGAATTTGATACACACTGTATT
- Phenotype
-
MIM: 604860
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MALT1
PubMed Links
Gene Details
- Gene
- MALT1
- Gene Name
- MALT1 paracaspase
View Full Product Details