Product Details

SNP ID
rs7576719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227325435 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTGTGGGCCCAGGGGTGAGTG[C/T]TGACGTCTTCCCGACCCGCGACCTG
Phenotype
MIM: 614785
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC654841 PubMed Links

Gene Details

Gene
LOC654841
Gene Name
uncharacterized LOC654841
There are no transcripts associated with this gene.

Gene
MFF
Gene Name
mitochondrial fission factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277061.1 154 Intron NP_001263990.1
NM_001277062.1 154 Intron NP_001263991.1
NM_001277063.1 154 Intron NP_001263992.1
NM_001277064.1 154 Intron NP_001263993.1
NM_001277065.1 154 Intron NP_001263994.1
NM_001277066.1 154 Intron NP_001263995.1
NM_001277067.1 154 Intron NP_001263996.1
NM_001277068.1 154 Intron NP_001263997.1
NM_020194.5 154 Intron NP_064579.3
XM_005246681.3 154 Intron XP_005246738.1
XM_005246682.1 154 Intron XP_005246739.1
XM_005246683.1 154 Intron XP_005246740.1
XM_005246684.1 154 Intron XP_005246741.1
XM_005246685.3 154 Intron XP_005246742.1
XM_005246686.3 154 Intron XP_005246743.1
XM_005246687.3 154 Intron XP_005246744.1
XM_005246688.1 154 Intron XP_005246745.1
XM_006712637.1 154 Intron XP_006712700.1
XM_006712638.1 154 Intron XP_006712701.1
XM_006712639.2 154 Intron XP_006712702.1
XM_011511500.1 154 Intron XP_011509802.1
XM_011511501.1 154 Intron XP_011509803.1
XM_017004512.1 154 UTR 5 XP_016860001.1
XM_017004513.1 154 Intron XP_016860002.1
XM_017004514.1 154 Intron XP_016860003.1
XM_017004515.1 154 Intron XP_016860004.1

View Full Product Details