Product Details
- SNP ID
-
rs6824288
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:84995876 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAATATTTCCAGGGGGATCATCAT[C/G]ATCCTCCCTCTCCTTTAGGAGCTAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDFY3-AS2
PubMed Links
Gene Details
- Gene
- WDFY3-AS2
- Gene Name
- WDFY3 antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details